(1R,2R)-trans-2-Aminocyclohexanol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-2-Aminocyclohexanol hydrochloride

A320048-1G A320048-100G A320048-10G A320048-250MG A320048-5G
103825-226EA 6 USD
103825-226 103836-158 76839-022 77154-484 77154-486 77154-488 77306-206
(1R,2R)-trans-2-Aminocyclohexanol hydrochloride 97%
(1R,2R)-trans-2-Aminocyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
Flash Pt: 99.7 °C
Storage Temperature: Ambient
MDL Number: MFCD09259962
CAS Number: 13374-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR