(1R,2R)-(+)-1,2-Diphenylethylenediamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1R,2R)-(+)-1,2-Diphenylethylenediamine, (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, (1R,2R)-1,2-Diphenylethane-1,2-diamine, (R,R)-DPEN, (+)-((R,R)-1,2-Diamino-2-phenylethyl)benzene
A139274-100G
A139274-10G
A139274-500G
A139274-1G
A139274-25G
A139274-5G
103802-722EA
5
USD
103802-722
76782-852
76782-854
76782-856
77299-068
77315-804
77315-806
(1R,2R)-(+)-1,2-Diphenylethylenediamine 98%
(1R,2R)-(+)-1,2-Diphenylethylenediamine
Precaución: for Research Use Only
Formula:
C₁₄H₁₆N₂ MW: 212.29 g/mol Boiling Pt: 353.9 °C (760 mm Hg) Melting Pt: 81…84 °C Flash Pt: 199.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082769 CAS Number: 35132-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...