(R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
A100110-100MG
A100110-250MG
A100110-5G
103795-256EA
59
USD
103795-256
103820-976
76970-110
76970-112
77282-432
(R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98%
(R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only
Formula:
C₁₅H₁₅N MW: 209.29 g/mol Boiling Pt: 338.4 °C (760 mm Hg) Melting Pt: 78…79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11858641 CAS Number: 180272-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...