(1R)-(+)-2,10,Camphorsultam 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ticagrelor Impurity 113, (3aR)-8,8-dimethylhexahydro-2H-3a,6-methanobenzo[d]isothiazole 1,1-dioxide, (3aR,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide
A364515-25G
A364515-5G
A364515-100G
A364515-10G
A364515-1G
103826-360EA
9
USD
103826-360
103837-378
76840-608
76840-610
77003-434
77003-436
77306-754
(1R)-(+)-2,10,Camphorsultam 95%
(1R)-(+)-2,10,Camphorsultam
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₂S MW: 215.32 g/mol Melting Pt: 185…187 °C Flash Pt: 150.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151762 CAS Number: 108448-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...