(1R)-(-)-(10-Camphorsulfonyl)oxaziridine 97%

Supplier: Ambeed
Danger

Synonyms: 1R)-(-)-(Camphorylsulphonyl)oxaziridine, (1R)-(-)-2,N-Epoxy-exo-10,2-bornanesultam

A163833-10G A163833-1G A163833-25G A163833-5G
103805-774EA 6 USD
103805-774 103844-038 76689-926 77320-156 77320-158 77320-160
(1R)-(-)-(10-Camphorsulfonyl)oxaziridine 97%
(1R)-(-)-(10-Camphorsulfonyl)oxaziridine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₃S
MW: 229.3 g/mol
Boiling Pt: 317.6 °C (760 mm Hg)
Melting Pt: 170…174 °C
Flash Pt: 145.9 °C
Storage Temperature: Freezer
MDL Number: MFCD30486441
CAS Number: 104372-31-8
UN: 1479
ADR: 5.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR