(1-Pyridin-4-yl)piperidine 97%

Supplier: Ambeed
Warning

A327960-100G A327960-10G A327960-500G A327960-25G A327960-5G
103836-434EA 8 USD
103836-434 77002-982 77002-984 77002-986 77306-362 77306-364
(1-Pyridin-4-yl)piperidine 97%
(1-Pyridin-4-yl)piperidine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂
MW: 162.2300 g/mol
Melting Pt: 78…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00006476
CAS Number: 2767-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR