1-Phenylpyrazole-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1-Phenyl-1H-pyrazol-4-yl)boronic acid

A170628-100MG A170628-250MG
103806-518EA 80 USD
103806-518 103844-792 76824-486 76824-488
1-Phenylpyrazole-4-boronic acid 98%
1-Phenylpyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₂O₂
Boiling Pt: 393.1±34.0 °C (760 mm Hg)
Flash Pt: 191.6 °C
Storage Temperature: Freezer
MDL Number: MFCD11044914
CAS Number: 1201643-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR