(1-Methylcyclopropyl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A139252-250MG A139252-1G
103802-718EA 150 USD
103802-718 77162-248 77299-064
(1-Methylcyclopropyl)methanamine hydrochloride 95%
(1-Methylcyclopropyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClN
MW: 121.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12962560
CAS Number: 1260779-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR