(1-Methyl-5-phenyl-1H-pyrazol-4-yl)boronic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A191275-100MG A191275-1G A191275-250MG
103816-360EA 451 USD
103816-360 103846-962 77683-500 77892-232 77892-233
(1-Methyl-5-phenyl-1H-pyrazol-4-yl)boronic acid ≥95%, Ambeed.Inc
(1-Methyl-5-phenyl-1H-pyrazol-4-yl)boronic acid

Precaución: For research use only.

Formula: C₁₀H₁₁BN₂O₂
Boiling Pt: 431.2±47.0 °C (760 mm Hg)
Flash Pt: 214.6 °C
Storage Temperature: Freezer
MDL Number: MFCD28040466
CAS Number: 1858252-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR