(1-Methyl-2-oxo-1,2-dihydropyridin-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A245859-100MG A245859-10G A245859-250MG A245859-1G A245859-5G
103811-676EA 41 USD
103811-676 103851-504 76987-764 76987-766 76987-768 77090-776 77090-778
(1-Methyl-2-oxo-1,2-dihydropyridin-4-yl)boronic acid 95%
(1-Methyl-2-oxo-1,2-dihydropyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
Storage Temperature: Freezer
MDL Number: MFCD22418629
CAS Number: 1351413-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR