1-Methyl-1H-pyrazole-5-boronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1-Methyl-1H-pyrazol-5-yl)boronic acid, (1-Methyl-1H-pyrazol-5-yl)-boronic acid
A201184-100G
A201184-10G
A201184-5G
103817-276EA
16
USD
103817-276
103847-914
77030-138
77030-140
77085-986
1-Methyl-1H-pyrazole-5-boronic acid 95%
1-Methyl-1H-pyrazole-5-boronic acid
Precaución: for Research Use Only
Formula:
C₄H₇BN₂O₂ Boiling Pt: 323 °C (760 mm Hg) Flash Pt: 149.1 °C Storage Temperature: Freezer |
MDL Number:
MFCD11053031 CAS Number: 720702-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...