(1-Methyl-1H-pyrazol-4-yl)methylamine 97%

Supplier: Ambeed
Warning

Synonyms: (1-Methyl-1H-pyrazol-4-yl)methanamine

A168712-5G A168712-10G A168712-250MG A168712-1G A168712-25G
103806-312EA 30 USD
103806-312 103844-566 77081-626 77163-428 77163-430 77300-938 77300-940
(1-Methyl-1H-pyrazol-4-yl)methylamine 97%
(1-Methyl-1H-pyrazol-4-yl)methylamine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.15 g/mol
Boiling Pt: 219.8 °C (760 mm Hg)
Flash Pt: 86.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02090924
CAS Number: 400877-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR