(1-Methyl-1H-indol-5-yl)methanamine 98%
Supplier: Ambeed
|
Danger
|
A621767-100MG
A621767-250MG
A621767-1G
A621767-5G
103840-220EA
404
USD
103840-220
76980-624
76980-626
77307-976
77307-978
(1-Methyl-1H-indol-5-yl)methanamine 98%
(1-Methyl-1H-indol-5-yl)methanamine
Precaución: for Research Use Only
Formula:
C₁₀H₁₂N₂ MW: 160.22 g/mol Melting Pt: 55 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09025839 CAS Number: 884507-17-9 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...