(1-Methyl-1H-imidazol-2-yl)methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-2-imidazolemethanol
A215005-100G
A215005-10G
A215005-25G
A215005-5G
103849-148EA
9
USD
103849-148
76999-752
76999-754
77087-674
77087-676
(1-Methyl-1H-imidazol-2-yl)methanol 97%
(1-Methyl-1H-imidazol-2-yl)methanol
Precaución: for Research Use Only
Formula:
C₅H₈N₂O MW: 112.13 g/mol Boiling Pt: 279.9 °C (760 mm Hg) Melting Pt: 116…119 °C Flash Pt: 123.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00964673 CAS Number: 17334-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...