(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol 97%

Supplier: Ambeed
Warning

A402587-1G A402587-25G A402587-5G A402587-100MG A402587-10G A402587-250MG
103827-078EA 14 USD
103827-078 103838-116 76841-578 76841-580 76841-582 76979-078 76979-080 76979-082
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol 97%
(1-Methyl-1H-[1,2,4]triazol-3-yl)methanol

Precaución: For research use only

Formula: C₄H₇N₃O
Boiling Pt: 296.8 °C (760 mm Hg)
Flash Pt: 133.329 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16710296
CAS Number: 135242-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR