(1H-Pyrazol-4-yl)methanol 97%

Supplier: Ambeed
Warning

A362770-1G A362770-25G A362770-5G A362770-100MG A362770-250MG
103826-330EA 39 USD
103826-330 103837-346 76840-560 76840-562 76840-564 77142-336 77142-338
(1H-Pyrazol-4-yl)methanol 97%
(1H-Pyrazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₆N₂O
Boiling Pt: 326.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06797673
CAS Number: 25222-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR