1H-Pyrazole-3-boronic acid 97%

Supplier: Ambeed
Warning

A103170-10G A103170-250MG A103170-25G A103170-1G A103170-5G A103170-100MG
103796-496EA 8 USD
103796-496 103822-200 77160-316 77160-318 77160-320 77309-074 77309-076 77891-761
1H-Pyrazole-3-boronic acid 97%
1H-Pyrazole-3-boronic acid

Precaución: for Research Use Only

Formula: C₃H₅BN₂O₂
Melting Pt: 85…88 °C
Storage Temperature: Freezer
MDL Number: MFCD02020768
CAS Number: 376584-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR