(1H-Indol-5-yl)methanamine 95%

Supplier: Ambeed
Warning

Synonyms: (1H-Indol-5-yl)methylamine, 5-(Aminomethyl)indole, (1H-Indol-5-yl)methanamine, Indole-5-methanamine

A115315-10G A115315-250MG A115315-1G A115315-5G
103798-544EA 26 USD
103798-544 103824-398 77160-960 77160-962 77297-362 77297-364
(1H-Indol-5-yl)methanamine 95%
(1H-Indol-5-yl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.189 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01719220
CAS Number: 81881-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR