5-Indoleboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid

A293610-5G A293610-10G A293610-1G A293610-25G
103813-986EA 14 USD
103813-986 103835-126 76837-746 77239-100 77305-638 77305-640
5-Indoleboronic acid 97%
5-Indoleboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
Flash Pt: 215.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319013
CAS Number: 144104-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR