(1H-Indol-3-yl)methanamine 95%

Supplier: Ambeed
Warning

Synonyms: Indole-3-methanamine

A118884-100MG A118884-10G A118884-250MG A118884-5G A118884-1G
103799-122EA 56 USD
103799-122 77136-404 77136-406 77136-408 77297-632 77311-794
(1H-Indol-3-yl)methanamine 95%
(1H-Indol-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.18 g/mol
Boiling Pt: 335.6 °C (760 mm Hg)
Melting Pt: 101…102 °C
Flash Pt: 183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631210
CAS Number: 22259-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR