(1H-Indol-3-yl)-1-propanamine 95%

Supplier: Ambeed
Warning

A100939-100MG A100939-250MG A100939-1G
103795-712EA 168 USD
103795-712 103821-400 77025-160 77025-162 77296-918
(1H-Indol-3-yl)-1-propanamine 95%
(1H-Indol-3-yl)-1-propanamine

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂
Flash Pt: 194.877 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130194
CAS Number: 6245-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR