(1H-Indazol-5-yl)methanol 98%
Supplier: Ambeed
|
Warning
|
A452117-100MG
A452117-250MG
A452117-1G
A452117-5G
103827-796EA
30
USD
103827-796
103838-866
76774-132
76774-134
76842-696
76842-698
(1H-Indazol-5-yl)methanol 98%
(1H-Indazol-5-yl)methanol
Precaución: for Research Use Only
Formula:
C₈H₈N₂O Flash Pt: 183.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038636 CAS Number: 478828-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...