1H-Indazole-4-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1H-Indazol-4-yl)boronic acid, 1H-indazole-4-boronic acid

A103451-25G A103451-1G A103451-5G A103451-250MG
103796-552EA 15 USD
103796-552 103822-258 77025-276 77309-128 77309-130 77599-506
1H-Indazole-4-boronic acid 97%
1H-Indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
MW: 161.9500 g/mol
Boiling Pt: 457 °C (760 mm Hg)
Flash Pt: 230.2 °C
Storage Temperature: Freezer
MDL Number: MFCD09878901
CAS Number: 1023595-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR