(1H-Imidazol-4-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A241837-100MG A241837-250MG
103811-432EA 106 USD
103811-432 103851-238 76815-420 76815-422
(1H-Imidazol-4-yl)methanamine hydrochloride 97%
(1H-Imidazol-4-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClN₃
MW: 133.5800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13176517
CAS Number: 66247-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR