1H-Imidazol-2-ylmethanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)-1H-imidazole, (1H-Imidazol-2-yl)methanol

A207173-1G A207173-250MG A207173-5G A207173-100MG
103817-766EA 30 USD
103817-766 77086-716 77139-548 77139-550 77685-538
1H-Imidazol-2-ylmethanol 97%
1H-Imidazol-2-ylmethanol

Precaución: for Research Use Only

Formula: C₄H₆N₂O
MW: 98.11 g/mol
Boiling Pt: 388.1 °C (760 mm Hg)
Flash Pt: 188.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06202766
CAS Number: 3724-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR