(1H-Benzoimidazol-2-yl)methanol 97%

Supplier: Ambeed
Warning

A101376-100G A101376-1G A101376-5G A101376-10G A101376-25G
103795-938EA 5 USD
103795-938 103821-624 77245-118 77297-072 77297-074 77570-098 77570-100
(1H-Benzoimidazol-2-yl)methanol 97%
(1H-Benzoimidazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 412.6 °C (760 mm Hg)
Melting Pt: 171…175 °C
Flash Pt: 203.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00014560
CAS Number: 4856-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR