(1-Benzyl-4-piperidinyl)methanol 97%
Supplier: Ambeed
|
Warning
|
A115612-10G
A115612-25G
A115612-1G
A115612-5G
103798-596EA
6
USD
103798-596
103824-454
76718-292
76718-294
77311-272
77311-274
(1-Benzyl-4-piperidinyl)methanol 97%
(1-Benzyl-4-piperidinyl)methanol
Precaución: for Research Use Only
Formula:
C₁₃H₁₉NO Boiling Pt: 312.2 °C (760 mm Hg) Flash Pt: 139.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00831014 CAS Number: 67686-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...