(1-Aminocyclopropyl)methanol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-1-(hydroxymethyl)cyclopropane hydrochloride, (1-Amino-cyclopropyl)-methanol hydrochloride

A115327-25G A115327-1G A115327-5G A115327-10G A115327-250MG
103798-546EA 11 USD
103798-546 103824-400 77026-050 77311-210 77311-212 77670-612 77670-614
(1-Aminocyclopropyl)methanol hydrochloride 97%
(1-Aminocyclopropyl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.5813 g/mol
Boiling Pt: 188.2 °C (760 mm Hg)
Melting Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD09834330
CAS Number: 115652-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR