(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

A277912-100G A277912-10G A277912-250MG A277912-1G A277912-25G A277912-5G
103813-382EA 19 USD
103813-382 103853-320 77032-112 77032-114 77032-116 77093-224 77305-280 77305-282
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 97%
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

Precaución: for Research Use Only

Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
Boiling Pt: 552.915 °C (760 mm Hg)
Melting Pt: 320 °C
Flash Pt: 288.193 °C
Storage Temperature: Ambient
MDL Number: MFCD00010045
CAS Number: 35193-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR