(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A129625-100MG A129625-250MG A129625-5G
103799-334EA 26 USD
103799-334 103801-304 77136-862 77136-864 77549-538
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 97%
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₁H₅₀F₆IrNP₂
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00075097
CAS Number: 64536-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR