1,4-Dioxan-2-ylmethanol 97%

Supplier: Ambeed
Warning

Synonyms: (1,4-Dioxan-2-yl)methanol

A538679-250MG A538679-1G A538679-5G
103828-614EA 35 USD
103828-614 103839-736 76818-986 76843-778 76843-780
1,4-Dioxan-2-ylmethanol 97%
1,4-Dioxan-2-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 208.2 °C (760 mm Hg)
Flash Pt: 96.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669844
CAS Number: 29908-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR