(1,3-Dioxolan-2-yl)methanol 95%

Supplier: Ambeed
Warning

A220670-1G A220670-25G A220670-5G A220670-10G A220670-250MG
103818-870EA 24 USD
103818-870 103849-752 77088-344 77088-346 77088-348 77165-196 77165-198
(1,3-Dioxolan-2-yl)methanol 95%
(1,3-Dioxolan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.11 g/mol
Flash Pt: 76.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062572
CAS Number: 5694-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR