1,3-Dimethyl-1H-pyrazole-5-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1,3-Dimethyl-1H-pyrazol-5-yl)boronic acid

A322975-1G A322975-250MG
103825-318EA 35 USD
103825-318 103836-256 76839-172 76875-888
1,3-Dimethyl-1H-pyrazole-5-boronic acid 95%
1,3-Dimethyl-1H-pyrazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₅H₉BN₂O₂
MW: 139.9500 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12407383
CAS Number: 847818-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR