(1,3-Dimethyl-1H-pyrazol-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A225574-100MG A225574-250MG A225574-1G
103819-186EA 56 USD
103819-186 103850-086 77013-388 77013-390 77088-788
(1,3-Dimethyl-1H-pyrazol-4-yl)boronic acid 97%
(1,3-Dimethyl-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₉BN₂O₂
MW: 139.95 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18250638
CAS Number: 1146616-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR