1,3,5-Trimethyl-1H-pyrazole-4-boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (1,3,5-Trimethyl-1H-pyrazol-4-yl)boronic acid

A127736-250MG A127736-1G A127736-5G
103801-004EA 39 USD
103801-004 76811-008 77313-268 77313-270
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid 96%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₆H₁₁BN₂O₂
MW: 153.9800 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11183310
CAS Number: 847818-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR