(1-(tert-Butoxycarbonyl)-5-((tert-butoxycarbonyl)amino)-1H-indol-2-yl)boronic acid 95%

Supplier: Ambeed
Warning

A100229-100MG A100229-250MG A100229-1G
103795-310EA 370 USD
103795-310 76821-192 76821-194 77282-462
(1-(tert-Butoxycarbonyl)-5-((tert-butoxycarbonyl)amino)-1H-indol-2-yl)boronic acid 95%
(1-(tert-Butoxycarbonyl)-5-((tert-butoxycarbonyl)amino)-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₅BN₂O₆
MW: 376.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11616305
CAS Number: 913388-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR