(1-(tert-Butoxycarbonyl)-4-fluoro-1H-indol-2-yl)boronic acid 97%

Supplier: Ambeed
Warning

A101983-1G A101983-250MG A101983-100MG
103796-236EA 105 USD
103796-236 103821-932 77308-814 77592-076 77670-468
(1-(tert-Butoxycarbonyl)-4-fluoro-1H-indol-2-yl)boronic acid 97%
(1-(tert-Butoxycarbonyl)-4-fluoro-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₄
MW: 279.07 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11109421
CAS Number: 1000068-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR