(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-1H-pyrazole-4-boronic acid, 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid, 1H-Pyrazole-4-boronic acid, N1-BOC protected

A195288-1G A195288-5G A195288-10G A195288-250MG
103816-736EA 19 USD
103816-736 103847-350 77085-300 77085-302 77600-588 77871-972
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid 98%
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₃BN₂O₄
Boiling Pt: 374.1 °C (760 mm Hg)
Melting Pt: 157…161 °C
Storage Temperature: Freezer
MDL Number: MFCD09951915
CAS Number: 1188405-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR