1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridine-4-boronic acid, (1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridin-4-yl)boronic acid
A159523-1G
A159523-250MG
A159523-100MG
A159523-10G
103805-264EA
87
USD
103805-264
103843-484
77300-406
77567-320
77671-260
77671-262
1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected 97%
1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected
Precaución: For research use only.
Formula:
C₁₀H₁₈BNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD09997795 CAS Number: 844501-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...