1-(Phenylsulfonyl)-3-indoleboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1-(Phenylsulphonyl)-1H-indol-3-yl)boronic acid

A342567-10G A342567-250MG A342567-5G
103825-866EA 75 USD
103825-866 103836-840 76788-298 76788-300 76839-884
1-(Phenylsulfonyl)-3-indoleboronic acid 95%
1-(Phenylsulphonyl)-3-indoleboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BNO₄S
MW: 301.1300 g/mol
Boiling Pt: 581.384 °C (760 mm Hg)
Flash Pt: 305.41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02681892
CAS Number: 129271-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR