(1-(2-Methoxyethyl)-1H-pyrazol-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A292600-1G A292600-250MG
103813-946EA 135 USD
103813-946 103835-086 76837-690 77571-804
(1-(2-Methoxyethyl)-1H-pyrazol-4-yl)boronic acid 95%
(1-(2-Methoxyethyl)-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₁₁BN₂O₃
MW: 169.97 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12407377
CAS Number: 847818-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR