pH and Ion Meters Accessories, Cole-Parmer
Supplier: Antyila Scientific
35816-71 35816-72
76221-314EA 227.91 USD
76221-314 76221-316
pH and Ion Meters Accessories, Cole-Parmer
Electrodes pH Electrodes
Get accurate readings for even demanding applications.
  • Choose a pH electrode based on your application
  • For clean samples, measure pH with single-junction electrodes
  • Use double-junction electrodes for samples with contaminations or particulates, heavy metals, or organic compounds
Low-maintenance epoxy-bodied electrodes provide the durability for field work. Refillable electrodes provide greater accuracy and faster response for more critical analytical work.