CRITERION™ LB Broth, Miller
Supplier: HARDY DIAGNOSTICS
C9272 C9271
76377-544EA 553.4 USD
76377-544 76446-886
CRITERION™ LB Broth, Miller
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ LB Broth, Miller is recommended for the cultivation and maintenance of Escherichia coli used in molecular biology procedures.
  • Wide mouth opening reduces dust formation by allowing the easy entry of a scoop
  • Stackable containers for efficient storage
  • Increased solubility dissolves quickly
  • Custom and bulk products available to your specifications
  • Complete technical/instruction sheets available in HUGO software
  • Unparalleled performance tested for optimal growth
Dehydrated culture media is a raw material not intended for use in the diagnosis of human disease. For implementation, this product requires additional processing and supplementation of ingredients before use.