Fargesin 97%

Supplier: Ambeed
Warning

Synonyms: rel-5-((1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl)benzo[d][1,3]dioxole, rel-5-[(1R,3aS,4S,6aS)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole

A185997-1MG A185997-5MG A185997-10MG
103791-542EA 68 USD
103791-542 103792-338 103792-340 76799-874 77084-124 77302-006
Fargesin 97%
Fargesin

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₆
MW: 370.40 g/mol
Boiling Pt: 506.388 °C (760 mm Hg)
Flash Pt: 208.923 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781423
CAS Number: 31008-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR