Pinocembrin 99%

Supplier: Ambeed
Warning

Synonyms: Galangin flavanone, (+)-Pinocoembrin, (+)-Pinocembrin, Dihydrochrysin, NSC 279005, (S)-5,7-Dihydroxy-2-Phenylchroman-4-One

A700854-100MG A700854-1MG A700854-10MG A700854-25MG A700854-5MG A700854-50MG
103791-716EA 23 USD
103791-716 103792-814 103792-816 77158-422 77158-424 77308-150 77308-152 77308-154 77686-685
Pinocembrin 99%
Pinocembrin

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₄
MW: 256.25 g/mol
Boiling Pt: 511.1 °C (760 mm Hg)
Flash Pt: 199.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06858345
CAS Number: 480-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR