(+)-Catechin hydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(+)-Catechin, (+)-3,3',4',5,7-Flavanpentol hydrate, (2R,3S)-2-(3,4-Dihydroxyphenyl)chromane-3,5,7-triol
A499582-1G
A499582-100MG
A499582-10MG
A499582-250MG
A499582-25G
A499582-50MG
A499582-5G
103828-278EA
36
USD
103828-278
76843-334
77116-552
77116-554
77116-556
77116-558
77116-560
77686-308
(+)-Catechin hydrate 97%
(+)-Catechin hydrate
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₆·xH₂O MW: 290.27 g/mol (anhy) Boiling Pt: 630.4 °C (760 mm Hg) Melting Pt: 175…177 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075649 CAS Number: 154-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...