Parthenolide 97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 4,5alpha-Epoxy-6beta-hydroxygermacra-1(10),11(13)-dien-12-oic Acid gamma-Lactone, (3aS,9aR,10aR,10bS,E)-6,9a-Dimethyl-3-methylene-3a,4,5,8,9,9a,10a,10b-octahydrooxireno[2',3':9,10]cyclodeca[1,2-b]furan-2(3H)-one

A105353-100MG A105353-1MG A105353-250MG A105353-1G
103796-876EA 146 USD
103796-876 103799-266 77245-386 77245-388 77245-390 77309-474
Parthenolide 97% (by HPLC)
Parthenolide

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₃
MW: 248.32 g/mol
Melting Pt: 115…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134592
CAS Number: 20554-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR