((4-Iodophenyl)ethynyl)trimethylsilane 97%

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-4-[2-(trimethylsilyl)ethynyl]-benzene

A283090-250MG A283090-1G A283090-25G A283090-5G
103813-602EA 54 USD
103813-602 103853-554 77141-206 77305-438 77305-440 77305-442
((4-Iodophenyl)ethynyl)trimethylsilane 97%
((4-Iodophenyl)ethynyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₃ISi
MW: 300.214 g/mol
Melting Pt: 66…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974026
CAS Number: 134856-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR