(4-Bromophenylethynyl)trimethylsilane 97%

Supplier: Ambeed
Warning

Synonyms: ((4-Bromophenyl)ethynyl)trimethylsilane

A280192-1G A280192-25G A280192-5G A280192-100G A280192-10G A280192-250MG
103813-498EA 8 USD
103813-498 103853-442 77093-374 77093-376 77093-378 77141-140 77141-142 77141-144
(4-Bromophenylethynyl)trimethylsilane 97%
(4-Bromophenylethynyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrSi
Boiling Pt: 247.1 °C (760 mm Hg)
Melting Pt: 61…63 °C
Flash Pt: 144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863556
CAS Number: 16116-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR