(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) 95%

Supplier: Ambeed
Warning

Synonyms: ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate, (7-Azabenzotriazol-1-yl)-N-oxy-tris(dimethyamino)phosphonium hexafluorosphate, AOP, Tris(dimethylamino)(3H-1,2,3-triazolo[4,5-b]pyridin-3-yloxy)phosphonium Hexafluorophosphate

A223666-100G A223666-10G A223666-1G A223666-25G A223666-5G
103819-054EA 14 USD
103819-054 103849-934 76691-872 76691-874 77303-660 77303-662 77303-664
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) 95%
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )

Precaución: for Research Use Only

Formula: C₁₂H₂₂F₆N₆OP₂
MW: 443.27 g/mol
Melting Pt: 180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09263289
CAS Number: 156311-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR